Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ BioCoat™ Collagen I Multiwell Plates
GREENER_CHOICE

Catalog No. 087742 Shop All Corning Products
Change view
Click to view available options
Culture Area:
3.8 cm2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Culture Area
08-774-2 3.8 cm2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 08-774-2 Supplier Corning™ Supplier No. 354500
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Treated with BioCoat™ for enhanced cell attachment and growth of a variety of primary cells and ransformed cell lines in serum-free or reduced serum cultures. Corning™ BioCoat™ Collagen I Multiwell Plates closely mimmics in vivo environments for cell culture applications. Ideal for applications involving adhesion, growth and differentiation of neuronal and transfected cell lines.

Collagen I, found in most tissues and organs, is most plentiful in dermis, tendon, and bone. It is an integral part of the framework that holds cells and tissues together and has been recognized as a useful matrix for improving cell culture. in vitro use of collagen can exert effects on the adherence, morphology, growth, migration, and differentiation of a variety of cell types.

Applications include:

  • Promotion of cell attachment and spreading
  • Rapid expansion of cell populations
  • Serum-free or reduced serum culture
  • Cell adhesion assays
  • Improving survival of primary cells in culture

Source

  • Rat tail tendon
  • Quality Control

    • Tested for ability to promote attachment and spreading of HT-1080 human fibrosarcoma cells
    • Tested and found negative for bacteria and fungi
    • Collagen I purity >90% by SDS-PAGE

    Storage

  • 4°C to 30°C under dry conditions.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Array 3 x 4
    Certifications/Compliance ANSI, SBS
    Closure Color Clear
    Closure Included Yes
    Closure Material Polystyrene
    Bottom Shape Flat
    Color Clear
    Culture Area 3.8 cm2
    Material Polystyrene
    Shape Rectangular
    Sterility Non-sterile
    Surface Treatment Cell Culture
    Type Multi-well Cell Culture Plate
    Well Count 12
    For Use With (Application) Promotion of cell attachment and spreading, rapid expansion of cell populations, serum-free or reduced serum culture, cell adhesion assay, improving survival of primary cells in culture
    Includes Lid
    No. per Case 5/Cs.
    No. per Pack 5/Pk.
    Quantity 5/Cs.
    Source Rat Tail
    Storage Requirements 4° to 30°C
    Show More Show Less
    WARNING: Cancer - www.P65Warnings.ca.gov
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.