Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ BioCoat™ 96-Well, Collagen Type IV-Treated, Flat-Bottom Microplate
GREENER_CHOICE

Catalog No. 0877430 Shop All Corning Products
Change view
Click to view available options
Color:
Clear
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
08-774-30 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 08-774-30 Supplier Corning™ Supplier No. 354429
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Color:Clear; Sterility:Sterile; Lid:With Lid

Corning™ BioCoatTM Collagen IV 96-well Clear Plate is comprised of clear TC-treated polystyrene with a uniform application of mouse collagen type IV. Flat bottom, with lid, 5/case

Corning BioCoat products come in a variety of pre-coated options and are an ideal solution for enhanced cell attachment and growth of a variety of primary cells and transformed cells in serum-free or serum-containing cultures. Corning offers custom coating capabilities.

TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Lid With Lid
Content And Storage 2°C to 8°C
Culture Area 0.32 cm2
Array 8 x 12
Color Clear
Product Type Microplate
Material Polystyrene
No. of Wells 96
Sterility Sterile
Surface Treatment Collagen Type IV
Well Shape Round, Flat Bottom (F-well)
For Use With (Application) Promotion of cell attachment and spreading, rapid expansion of cell populations, serum-free or reduced serum culture, cell adhesion assay, improving survival of primary cells in culture
Quantity 5/Cs.
Source EHS Lathrytic Mouse Tumor
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.