Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ BioCoat™ Fibronectin Multiwell Plate
GREENER_CHOICE

Catalog No. 0877456 Shop All Corning Products
Change view
Click to view available options
Culture Area:
9.6 cm2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Culture Area
08-774-56 9.6 cm2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 08-774-56 Supplier Corning™ Supplier No. 354402
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Promotion of cell attachment and spreading

Human Fibronectin (HFN) is a widely distributed glycoprotein that is used as a substrate to promote attachment of cells through its central-binding domain RGD sequence. HFN is a product of most mesenchymal and epithelial cells and is present in both the ECM and plasma. The principal function of HFN appears to be in cellular migration during wound healing and development, regulation of cell growth and differentiation, and haemostasis/thrombosis.

Applications include:

  • Promotion of cell attachment and spreading
  • Rapid expansion of cell populations
  • Serum-free or reduced-serum culture
  • Cell adhesion assays
  • Studies of effects of HFN on cell behavior
  • Improving survival of primary cells in culture

Source

  • Human plasma

  • NOTE: Source material tested for hepatitis B antigen and HIV-1 antibody

    Quality Control

    • Tested for ability to promote attachment and spreading of BHK-21 hamster kidney cells
    • Tested and found negative for bacteria and fungi
    • Fibronectin purity >90% by SDS-PAGE

    Storage

  • 2°C to 8°C. Do not freeze.
  • Order Info

    Supplied in ready-to-use Falcon™ Companion Tissue Culture Plates

    TRUSTED_SUSTAINABILITY

    Specifications

    Array 2 x 3
    Closure Color Clear
    Closure Included Yes
    Closure Material Polystyrene
    Bottom Shape Flat
    Color Clear
    Culture Area 9.6 cm2
    Material Polystyrene
    Shape Rectangular
    Sterility Non-sterile
    Surface Treatment Cell Culture
    Type Multi-well Cell Culture Plate
    Well Count 6
    For Use With (Application) Promotion of cell attachment and spreading, rapid expansion of cell populations, increased proliferation of PC12 cells, studies of effects of collagen IV on cell behavior, cell adhesion assays
    Includes Lid
    No. per Case 5/Cs.
    No. per Pack 1/Pk.
    Quantity 5/Cs.
    Source Human Fibronectin
    Storage Requirements 2°C to 8°C
    Height (Metric) 12 mm
    Diameter (Metric) 35 mm
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.