Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ BioCoat™ Intestinal Epithelium Differentiation Environment Kit

Catalog No. 08774175 Shop All Corning Products
Change view
Click to view available options
Intestinal Epithelium Differentiation Environment Kit; 1/Cs
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
08-774-175 Intestinal Epithelium Differentiation Environment Kit; 1/Cs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 08-774-175 Supplier Corning™ Supplier No. 355057

Includes

  • 400mL Entero-STIM™ Enterocyte Differentiation Medium (DMEM containing sodium butyrate)
  • Lyophilized BD™ MITO+ Serum Extender Supplement
  • 100mL Seeding Basal Medium (DMEM)
  • 24 ™ Fibrillar Collagen Cell Culture Inserts (inserts contain 1μm pore size membranes packaged in one 24-well Falcon™ Companion TC Plate with one additional plate

Used to create intestinal models that exhibit barrier function for use in transport and absorption studies of drugs, metabolites and infectious diseases of the intestine

Corning™ BioCoat™ Intestinal Epithelium Differentiation Environment is an integrated system designed to create in vitro intestinal models. The system comprises a specially formulated serum-free medium, culture supplements, sodium butyrate, and Corning BioCoat Fibrillar Collagen Cell Culture Inserts in one convenient kit.

For Customers in California only: WARNING Please refer to Safety Data Sheet (SDS) Section 15 or Letter in Lieu for more information regarding California Proposition 65.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Transport and absorption studies of drugs, metabolites and infectious diseases of the intestine
Storage Requirements 2° to 8°C
No. per Pack 1/Pk.
No. per Case 1/Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.