Learn More
Description
Corning BioCoat products come in a variety of pre-coated options and are an ideal solution for enhanced cell attachment and growth of a variety of primary cells and transformed cells in serum-free or serum-containing cultures. Corning offers custom coating capabilities.
Specifications
Specifications
| Barcode | No Barcode |
| Format | Standard |
| Lid | With Lid |
| Content And Storage | 2°C to 8°C |
| Culture Area | 0.32 cm2 |
| Array | 8 x 12 |
| Packaging | 1/Pk., 5/Cs. |
| Color | Clear |
| Product Type | Microplate |
| Material | Polystyrene |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
