Learn More
Corning™ BioCoat™ Matrigel™ Invasion Chamber: With GFR Matrigel Matrix
GSA_VAGREENER_CHOICE
Catalog No. 08774193 Shop All Corning Products
Change view
Click to view available options
Pore Size:
8 μm
Catalog No. Pore Size
08-774-193 8 μm
1 options
Catalog No. 08-774-193 Supplier Corning™ Supplier No. 354483
Add to Cart
Add to Cart
Provide rapid and reproducible quantification of cell invasion in vitro
Corning BioCoat cell culture inserts are pre-coated with extracellular matrix proteins for applications requiring a protein-coated cell surface, such as cell adhesion, growth, invasion, migration and/or differentiation.
Coatings include Corning Matrigel™ matrix, Fibronectin, Collagen, and Laminin.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Culture Area 0.3 cm2
Material PET
Pore Size 8 μm
Type Plate
For Use With (Application) Studies in which minimal interference by growth factors is desirable
Quantity 12/Pk., 24/Cs.
Surface Coating Matrigel™ Matrix
Inserts per Plate 12