Description
Corning™ BioCoat™ products are an ideal solution for enhanced cell attachment and growth of a variety of primary cells and transformed cells in serum-free or serum-containing cultures. Corning offers custom coating capabilities.
Shelf life: ≥ 3 months from date of shipment
Shipping room temperature.
Order Info
Bulk quantities available; please contact your Customer Service Representative.
Specifications
Specifications
| Bottom Shape | Round |
| Culture Area | 58.1 cm2 |
| Shape | Round |
| Surface Treatment | Cell Culture |
| Type | Cell Culture Dish |
| No. per Case | 10/Cs. |
| No. per Pack | 5/Pk. |
| Diameter (Metric) | 100 mm |