Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ BioCoat™ Poly-D-Lysine/Laminin Multiwell Plate
GREENER_CHOICE

Catalog No. 08774168 Shop All Corning Products
Change view
Click to view available options
Culture Area:
9.6 cm2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Culture Area
08-774-168 9.6 cm2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 08-774-168 Supplier Corning™ Supplier No. 354595
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Enhancement of neuronal cell attachment to plastic and glass

For some applications, the use of a combination of ECM proteins, such as Laminin (LM) and Fibronectin (HFN) or LM and attachment factors such as Poly-D-Lysine (PDL) or Poly-L-Orinthine (PLO) has been shown superior to the use of either alone. Corning BioCoat PDL/LM and PLO/LM Cultureware is suitable for culturing many different types of Peripheral Nervous System (PNS) and Central Nervous System (CNS) networks and is useful for promoting neural cell attachment and differentiation. Corning BioCoat LM/HFN Cultureware provides an in vitro environment that promotes cell attachment and extensive process formation.

Applications include:

  • Enhancement of neuronal cell attachment to plastic and glass
  • Promotion of neurite outgrowth
  • Culture of glial cells as a feeder layer for neurons
  • Construction of neural cell model systems to study CNS function, development, and diseases

Source

  • PDL, synthetic (MW 75-150 kD)
  • PLO, synthetic (MW 30-70 kD)
  • Laminin, EHS mouse tumor
  • Fibronectin, human plasma
  • NOTE: Source material tested for hepatitis B antigen and HIV-1 antibody.
  • Quality Control

    • PDL/LM and PLO/LM tested for ability to initiate differentiation (neurite outgrowth) of NG-108 rat glioma/mouse neuroblastoma cells
    • LM/HFN tested for receptor agonist induced changes in intracellular calcium using Fluo-3A (a long wavelength fluorescent calcium indicator) in primary rat cortical neuron enriched cultures
    • Tested and found negative for bacteria and fungi

    Storage

  • 2° to 8°C. Do not freeze.
TRUSTED_SUSTAINABILITY

Specifications

Array 2 x 3
Closure Color Clear
Closure Included Yes
Closure Material Polystyrene
Bottom Shape Flat
Color Clear
Culture Area 9.6 cm2
Material Polystyrene
Shape Rectangular
Sterility Non-sterile
Surface Treatment Poly-D-Lysine
Type Multi-well Cell Culture Plate
Well Count 6
For Use With (Application) Enhancement of neuronal cell attachment to plastic and glass, promotion of neurite outgrowth, culture of glial cells as a feeder layer for neurons, construction of neural cell model systems to study CNS function, development and diseases
Includes Lid
No. per Case 5/Cs.
No. per Pack 1/Pk.
Quantity 5/Cs.
Source Poly-D-Lysine and Laminin
Storage Requirements 2°C to 8°C
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.