Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ BioCoat™ 96-Well, Laminin/Poly-D Lysine-Treated, Flat-Bottom Microplate
GREENER_CHOICE

Catalog No. 08774170 Shop All Corning Products
Change view
Click to view available options
Color:
Clear
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
08-774-170 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 08-774-170 Supplier Corning™ Supplier No. 354596
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Color:Clear; Sterility:Sterile; Lid:With Lid

Corning™ BioCoatTM Poly-D-Lysine/Laminin 96-well Clear Plate is comprised of clear TC-treated polystyrene with a uniform application of Poly-D-Lysine and laminin. Flat bottom, with lid, 5/case

Corning BioCoat products come in a variety of pre-coated options and are an ideal solution for enhanced cell attachment and growth of a variety of primary cells and transformed cells in serum-free or serum-containing cultures. Corning offers custom coating capabilities.

TRUSTED_SUSTAINABILITY

Specifications

Format Standard
Lid With Lid
Content And Storage 2°C to 8°C
Culture Area 0.32 cm2
Array 8 x 12
Packaging 1/Pk., 5/Cs.
Color Clear
Product Type Microplate
Material Polystyrene
No. of Wells 96
Sterility Sterile
Surface Treatment Laminin/Poly-D-Lysine
Well Shape Flat Bottom (F-well)
For Use With (Application) Enhancement of neuronal cell attachment to plastic and glass, promotion of neurite outgrowth, culture of glial cells as a feeder layer for neurons, construction of neural cell model systems to study CNS function, development and diseases
Quantity 5/Cs.
Source Poly-D-Lysine and Laminin
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.