Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ BioCoat™ Poly-D-Lysine Multiwell Plates
GREENER_CHOICE

Catalog No. 08774280 Shop All Corning Products
Change view
Click to view available options
Well Count:
48
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Well Count
08-774-280 48
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 08-774-280 Supplier Corning™ Supplier No. 354509
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Designed to closely mimic in vivo environments and enhance expression of cell-specific morphology. Corning™ BioCoat™ Poly-D-Lysine (PDL) Multiwell Plates are ideal for applications involving adhesion, growth and differentiation of neuronal and transfected cell lines.

Poly-D-Lysine (PDL) and Poly-L-Lysine (PLL) are synthetic compounds that enhance cell adhesion and protein absorption by altering surface charges on the culture substrate. In addition to promoting cell adhesion, poly-lysine surface treatments support neurite outgrowth and improve the survival of many central nervous system (CNS) primary cells in culture. As PDL and PLL are synthetic molecules, they do not stimulate biological activity in the cells cultured on them, and they do not introduce impurities carried by natural polymers.

Applications include:

  • Attachment and spreading of a variety of cell types
  • Cell differentiation and neurite outgrowth
  • Attachment of fastidious transfected cell lines
  • Support survival of primary neurons in culture
  • Serum-free or reduced-serum culture

Source

  • PDL, synthetic (MW 75-150 kD)
  • PLL, synthetic (MW 30-70 kD)

Quality Control

  • Tested for ability to promote firm attachment of RCG cells
  • Tested and found negative for bacteria and fungi

Storage

  • 4°C to 30°C under dry conditions.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Array 6 x 8
    Certifications/Compliance ANSI, SBS
    Closure Color Clear
    Closure Included Yes
    Closure Material Polystyrene
    Bottom Shape Flat
    Color Clear
    Culture Area 0.75 cm2
    Material Polystyrene
    Shape Rectangular
    Sterility Non-sterile
    Surface Treatment Cell Culture
    Type Multi-well Cell Culture Plate
    Well Count 48
    For Use With (Application) Attachment and spreading of a variety of cell types, cell differentiation and neurite outgrowth, attachment of fastidious transfected cell lines, support survival of primary neurons in culture, serum-free or reduced serum culture
    Includes Lid
    No. per Case 5/Cs.
    No. per Pack 5/Pk.
    Quantity 5/Cs.
    Source Poly-D-Lysine
    Storage Requirements 4°C to 30°C
    Show More Show Less
    WARNING: Reproductive Harm - www.P65Warnings.ca.gov
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.