Learn More
Sigma Aldrich Fine Chemicals Biosciences Corning bottle-top vacuum filter system cellulose acetate membrane, pore size 0.22m, membrane area 54.5cm2, filter capacity 1000mL
Supplier:  Sigma Aldrich Fine Chemicals Biosciences CLS43051712EA
Encompass
Corning bottle-top vacuum filter system cellulose acetate membrane, pore size 0.22m, membrane area 54.5cm2, filter capacity 1000mL
Catalog No. 50-165-7435