Learn More
Corning™ Bovine Pituitary Extract (BPE)
GREENER_CHOICE
Catalog No. CB40123 Shop All Corning Products
Change view
Click to view available options
Quantity:
15 mg
Catalog No. Quantity
CB-40123 15 mg
1 options
Catalog No. CB-40123 Supplier Corning™ Supplier No. 354123
Add to Cart
Edge
Add to Cart
Supplement used in the culture of a variety of epithelial and endothelial cells
The Corning portfolio of cytokines, growth factors, and media supplementation offers a wide array of proteins that result in proliferation or differentiation of the affected cells.
  • Vialed, highly purified growth factors with high biological activity
  • Broad range of highly purified natural and recombinant lymphokines
  • Enriched culture supplements to be reconstituted/added in the medium of your choice
  • Propagate your cells under more defined serum-reduced or serum-free conditions
Corning offers a broad range of other media growth supplements. Some products are specific to cell type, such as SMC4 (for iPSCs) and T-Cell culture supplement (for T-cells), while other products promote proliferation in a wide variety of cell types. Reducing serum use is made easy with products such as ITS and Corning Nu-SerumTM.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage 2° to 8°C
Product Type Bovine Pituitary Extract
Quantity 15 mg