Learn More
Corning™ CellBIND™ 96-Well, CellBIND-Treated, Flat-Bottom Microplate
BIOPHARMAGSA_VAGREENER_CHOICE
Catalog No. 0720194 Shop All Corning Products
Change view
Click to view available options
Color:
Clear
Catalog No. Color
07-201-94 Clear
1 options
This item is not returnable. View return policy
Catalog No. 07-201-94 Supplier Corning™ Supplier No. 3300
  • PRODUCT PROMOTION
    $786.50
    $459.00 / Case of 50
    Save $327.50
    42% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy
Color:Clear; Sterility:Sterile; Lid:With Lid
  • Cell culture microplates are sterile and nonpyrogenic.
  • Lids available where indicated
Cat. nos. 3798 and 3898 processed to improve hydrophilicity for hemagglutination and similar assays.
Cat. Nos. 3585 and 3595 have special low evaporation lids.
Cat. No. 3841 is aseptically manufactured.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Barcode No Barcode
Lid With Lid
Culture Area 0.32 cm2
Array 8 x 12
Binding Type Corning CellBIND Surface
Color Clear
Product Type Microplate
Material Polystyrene
No. of Wells 96
Volume (Metric) Working 75 to 200 μL
Sterility Sterile
Surface Treatment CellBIND
Total Volume (Metric) Well 360 μL
Well Shape Flat Bottom (F-well)
For Use With (Application) CellBIND-treated surface helps cells adapt to reduced serum or serum-free media and improves cell attachment for increased cell growth and yield. Requires no refrigeration or special handling, unlike standard coatings used to improve cell attachment.
No. per Case 50/Cs.
No. per Pack 5/Pk.
Quantity 50/Cs.
Well Color Clear