Learn More
Corning™ CellCube™ Tubing Manifold
GREENER_CHOICE
Catalog No. 10320189 Shop All Corning Products
Change view
Click to view available options
Includes:
Thermoelastic Tubing, Cross Connection, 4 Aseptic Connectors
Catalog No. Includes
10-320-189 Thermoelastic Tubing, Cross Connection, 4 Aseptic Connectors
1 options
Catalog No. 10-320-189 Supplier Corning™ Supplier No. 3235
Add to Cart
Add to Cart
A new closed system offering designed to help reduce the risk of adventitious contamination
  • Closed System Cell Cube
  • CellCube 10, 25, 100 modules, circulation loops, and connectors
  • Allows for easy adherent cell scale-up that integrate seamlessly with AseptiQuik™ and MPC connectors.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Corning CellCube
Includes Thermoelastic Tubing, Cross Connection, 4 Aseptic Connectors
No. per Case 2/Cs.
No. per Pack 1/Pk.
Product Type Tubing Manifold
Sterility Sterile
Quantity 2 Pk.