Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ Cell Culture Media: Cell Culture Supplements
GSA_VA

Catalog No. p-3202715 Shop All Corning Products
Change view
Click to view available options
pH:
5.2 to 0.1
Neutral
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. pH
MT25037CI Neutral
MT-46033CI 5.2 to 0.1
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MT25037CI Supplier Corning™ Supplier No. 25037CI
Only null left
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Used as a nutrient source in cell culture media. Cell Culture Media: Cell Culture Supplements are produced under the highest industry standards to ensure superior results.

(This is suggested replacement for 25-037-CIR.)

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 15° to 30°C
Product Type Glucose
Sterility Sterile-filtered
pH Neutral
Quantity 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.