Learn More
Description
- Maintains cell culture media in the physiological pH range
- Maintains cell tonicity and viability
- Produced under the highest industry standards to ensure superior results
- Nontoxic
- Liquid without calcium and magnesium
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Concentration | 10 X |
| pH | 7.3 to 7.5 |
| Physical Form | Liquid |
| Quantity | 6 x 1000 mL |
| Solution Type | Buffer |
| Formulation | Potassium Chloride 2g/L, Potassium Dihydrogen Phosphate 2.40g/L, Sodium Chloride 80g/L, Disodium Phosphate 14.4g/L |
| Includes | No Calcium, No Magnesium |
| Grade | Not Available |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
