Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ Cell Culture Phosphate Buffered Saline (1X)
GSA_VA

Catalog No. MT21040CMX Shop All Corning Products
Change view
Click to view available options
Quantity:
6 x 500 mL
6 x 1000 mL
12 x 1000 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MT21040CMX 12 x 1000 mL
MT21040CV 6 x 500 mL
MT21040CM 6 x 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MT21040CMX Supplier Corning™ Supplier No. 21040CMX12
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

With low endotoxin content. Without calcium and magnesium.

Phosphate buffered saline (PBS) is a buffer solution containing sodium chloride, sodium phosphate, and in some formulations potassium chloride and potassium phosphate. PBS helps to maintain a constant pH and is isotonic and non-toxic to cells. This formulation does not contain calcium and magnesium.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 X
pH 7.3 to 7.5
Physical Form Liquid
Quantity 12 x 1000 mL
Formulation Potassium Dihydrogen Phosphate 0.144g/L, Sodium Chloride 9g/L, Disodium Phosphate 0.795g/L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.