Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ G418 Sulfate, 50mg/mL
GSA_VA

Catalog No. p-4877180 Shop All Corning Products
Change view
Click to view available options
Quantity:
20 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MT30234CR 20 mL
MT30234CI 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MT30234CR Supplier Corning™ Supplier No. 30234CR
Only null left
Add to Cart
Edge
Add to Cart

Blocks polypeptide synthesis by hindering protein elongation. Corning™ G418 Sulfate is an aminoglycoside similar in structure to gentamicin, neomycin and kanamycin.

Well-known, effective selection agent G418 Sulfate is recommended for use as selection agent in a concentration range of 100-800 μg/mL. This antibiotic blocks polypeptide synthesis by hindering protein elongation, and inhibits the selection and maintenance of eukaryotic cells stably transfected with the neomycin resistance gene.  G418 is an aminoglycoside similar in structure to gentamicin, neomycin and kanamycin.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2° to 8°C
Concentration 50 mg/ mL Solution (active G418)
Form Liquid
Product Type Antibiotic
Sterility Sterile
Format Plastic Bottle
Quantity 20 mL
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.