Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ Liquid Media in 10- and 20-Liter Bags

Catalog No. MT10040LX Shop All Corning Products
Change view
Click to view available options
Quantity:
10 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MT10040LX 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MT10040LX Supplier Corning™ Supplier No. 10040LX
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Flexible media-filled containers supply 10- and 20-liter quantities of media in a compact package for more efficient shipping, storage and handling

(This is suggested replacement for 10-040-LXR.)

Order Info

Media tote and collection containers available separately

TRUSTED_SUSTAINABILITY

Specifications

Product Type RPMI 1640
Quantity 10 L
pH 7.2 ±0.2
Content And Storage 2° to 8°C
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.