Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ Minimum Essential Medium Eagle (Mod.) 1X (MEM) without Glutamine
GSA_VA

Catalog No. p-4907452 Shop All Corning Products
Change view
Click to view available options
Quantity:
6 x 500 mL
6 x 1 L
Without Additives:
No Glutamine
No Glutamine, No Phenol Red
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Without Additives
MT15010CM 6 x 1 L No Glutamine
MT15010CV 6 x 500 mL No Glutamine
MT17305CV 6 x 500 mL No Glutamine, No Phenol Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MT15010CM Supplier Corning™ Supplier No. 15010CM
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Guaranteed low endotoxin levels ≤0.025EU/mL.

Minimum Essential Medium (MEM) is a common cell culture medium developed from early work using Basal Medium Eagle (BME) with normal mammalian fibroblasts and certain subtypes of HeLa cells.  This research indicated that additions made to BME aided cell propagation.  MEM is modified with higher concentrations of amino acids to more closely approximate the protein composition of mammalian cells.  When supplemented with serum, MEM has been used for cultivation of a wide variety of cells grown in monolayer, such as fibroblast.  This formulation contains sodium bicarbonate and does not contain L-glutamine.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Form Liquid
Product Type MEM Eagle/EBSS
Sterility Sterile
With Additives Earle's Balanced Salt Solution (EBSS)
Without Additives No Glutamine
Quantity 6 x 1 L
pH 7 to 7.4
Content And Storage 2° to 8°C
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.