Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ Penicillin-Streptomycin-L-Glutamine

Catalog No. MT30009CI Shop All Corning Products
Change view
Click to view available options
Quantity:
6 x 100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MT30009CI 6 x 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MT30009CI Supplier Corning™ Supplier No. 30009CI
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Packaged in plastic bottles

Technical Advantage:  Effective against both Gram-positive and -negative bacteria
Penicillin-Streptomycin-L-glutamine combines two antibiotics with a nutrient which is essential for cell growth, L-glutamine.  Penicillin inhibits bacterial growth by inhibiting peptidoglycan synthesis (essential for the bacterial cell wall), and streptomycin inhibits bacterial growth by inhibiting protein synthesis.  L-Glutamine is essential as an energy source for most mammalian cells in culture because of its amino acid carbon skeleton.  This antibiotic-nutrient combination is effective against Gram-negative and Gram-positive bacteria.
-  Complete Certificate of Analysis available for each production lot, along with full formulation information
-  Produced under the highest industry standards to ensure superior results
10,000 I.U. Penicillin (per mL) 10,000 μg/ mL Streptomycin
Corning media certificates are available on our Resource Library.

For Customers in California only: WARNING Please refer to Safety Data Sheet (SDS) Section 15 or Letter in Lieu for more information regarding California Proposition 65.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -20°C
Concentration 10,000 I.U. Penicillin (per mL); 10,000 ug/mL Streptomycin; 29.2 mg/mL L-glutamine
Form Liquid
Product Type Antibiotic-L-Glutamine
Format Plastic Bottle
Quantity 6 x 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.