Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris Base Buffer, Powder, Corning™

Catalog No. MT61233RR Shop All Corning Products
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MT61233RR 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MT61233RR Supplier Corning Supplier No. 61233RR
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used in buffer solutions in biochemical and molecular biological research

Cellgro incorporates many principles developed by the pharmaceutical industry including the use of WFI quality water, raw material testing, in-process monitoring, and finished product testing.

  • pH: 7.0 to 9.2
  • Molecular weight: 121.14

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tris Base Buffer
CAS 77-86-1
For Use With (Application) Molecular Biology
pH 7 to 9.2
Physical Form Powder
Quantity 1 kg
Solution Type Buffer
Packaging Plastic Bottle
Molecular Weight (g/mol) 121.14
Grade Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.