Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ Trypan Blue
BIOPHARMAGSA_VA

Catalog No. MT25900CI Shop All Corning Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MT25900CI 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MT25900CI Supplier Corning™ Supplier No. 25900CI
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Can be used for variety cell counting methods requiring information on cell count and viability.

Cellgro incorporates many principles developed by the pharmaceutical industry including the use of WFI quality water, raw material testing, in-process monitoring, and finished product testing.

  • 0.4% (w/v) solution
  • In PBS
  • pH 7.5 ±0.5
  • Quantity: 100mL
  • Viable cells exclude the dye, while nonviable cells absorb the dye and appear blue
  • Cells should be in suspension as single cells in buffered saline before counting
  • Can be used in assays providing total cell count, viable cell or nonviable cell count alone
  • Solution is provided at the common concentration of 0.4% (w/v)

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Solution Type Trypan Blue 0.4% Solution in PBS
Quantity 100 mL
Format Liquid
Storage Requirements 15° to 30°C
Volume (Metric) 100 mL
pH 7.5 ±0.5
Packaging Plastic Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.