Learn More
Corning™ CellSTACK™ Filling Accessory
BIOPHARMAGREENER_CHOICE
Catalog No. 15100126 Shop All Corning Products
Change view
Click to view available options
Sterility:
Sterile
Catalog No. Sterility
15-100-126 Sterile
1 options
This item is not returnable. View return policy
Catalog No. 15-100-126 Supplier Corning™ Supplier No. 11902
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy
Corning™ CellSTACK™ Filling Accessory, with Female MPC.
  • Sterile
  • Female MPC connector
  • C-Flex tubing with 9.5 mm (3/8 in.) inside diameter
  • 71.1 cm (28 in.) tubing length
TRUSTED_SUSTAINABILITY
Specifications
Closure Material Virgin HDPE
Description Corning™ CellSTACK™ Filling Accessory
For Use With (Equipment) Corning CellSTACK™ Culture Chambers
No. per Case 4/Cs.
No. per Pack 1/Pk.
Product Type Filling Accessory
Sterility Sterile
Closure Size 9.5 mm
Quantity 1 Ea.
Packaging Bag
WARNING: Cancer - www.P65Warnings.ca.gov