Learn More
Corning™ CellSTACK™ Stacker
Catalog No. 07201631 Shop All Corning Products
Change view
Click to view available options
Material:
Plastic
Catalog No. Material
07-201-631 Plastic
1 options
Catalog No. 07-201-631 Supplier Corning™ Supplier No. 3331
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart
Material: Plastic; For Use With: Incubator
A stacking device models the top surface of the CellSTACK™ chamber, securely nesting in the strengthening ribs. It is used to optimize incubator space while providing clearance for venting
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Incubator
Material Plastic
Type Stacker
For Use With (Application) Incubator
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov