Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ Collagen I, Rat
GSA_VAGREENER_CHOICE

Catalog No. p-4696757 Shop All Corning Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CB-40236 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. CB-40236 Supplier Corning™ Supplier No. 354236
Only null left
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Enhances cell attachment and proliferation or promotes expression of cell-specific morphology and function. Corning™ Collagen I, Rat is effective as a gel or a thin coating at a recommended concentration of 5 to 10μg/cm2 of growth surface depending on cell type.

Corning extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.

Corning offers a broad range of collagen types that are derived from multiple species. These collagens have been used to culture a variety of cell types to promote adhesion, growth, and/or differentiation.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 3 to 4 mg/mL
For Use With (Application) Cell Attachment and Proliferation, Cell-specific Morphology and Function
Formulation Liquid in 0.02N acetic acid
Quantity 100 mg
Source Rat Tail Tendon
Product Type Collagen, Rat Tail Type I
Content And Storage 2°C to 8°C
Purity or Quality Grade >90% by SDS-PAGE
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.