Learn More
Corning™ Costar™ Thermowell™ Plate Holder
GREENER_CHOICE
Catalog No. 0720021 Shop All Corning Products
Change view
Click to view available options
For Use With (Equipment):
Thermo well polycarbonate plates
Catalog No. For Use With (Equipment)
07-200-21 Thermo well polycarbonate plates
1 options
Catalog No. 07-200-21 Supplier Corning™ Supplier No. 2572
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart
For Thermo well polycarbonate plates
  • 96-well egg crate style holder for 1 x 8 Stripwell™ strips
  • Holds twelve 1 x 8 strips
  • Allows each individual well to be positioned back into the plate once broken apart
TRUSTED_SUSTAINABILITY
Specifications
Type Plate Holder
For Use With (Equipment) Thermo well polycarbonate plates