Learn More
Corning™ Costar™ Transtar-96™ 96-Well Liquid Transfer System
Catalog No. 0720059 Shop All Corning Products
Change view
Click to view available options
Channels:
96
Catalog No. Channels
07-200-59 96
1 options
This item is not returnable. View return policy
Catalog No. 07-200-59 Supplier Corning™ Supplier No. 7605
This item is not returnable. View return policy
Portable, autoclavable liquid handling system designed for use with 96 well plates.
The Costar™ Transtar-96™ System is a portable, autoclavable liquid handling system designed for use with 96 well plates. A sterile 96 tip disposable cartridge, which loads into the Transtar-96™ system, enables liquids to be aspirated, transferred and dispensed over a volume range of 25 to 200 μL in 5 μL increments. -Ideal for changing cell culture media and screening monoclonal antibodies. -Accuracy is rated at +/- 5% at all volume levels. -For sterile replacement cartridges, see No. 7610. -For sterile replacement reservoirs, see Nos. 4876 (open), 4877 (12-channel), or 4878 (8-channel). Refer to accessories tab below.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Liquid Transfer System
Autoclavable Autoclavable
Channels 96
For Use With (Application) Designed for use with 96-well plates, changing cell culture media and screening monoclonal antibodies
Sterility Sterile