Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ External Temperature Controller
SCI_ED

Catalog No. S90870TM Shop All Corning Products
Change view
Click to view available options
Temperature (Metric):
Ambient to 200°C
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Temperature (Metric)
S90870TM Ambient to 200°C
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
CAUTION: A 200°C setting when using 6795PR does not mean that the top plate is at 200°C. Top plate temperature could be as high as 550°C.
Catalog No. S90870TM Supplier Corning™ Supplier No. 6795PR/EMD

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
CAUTION: A 200°C setting when using 6795PR does not mean that the top plate is at 200°C. Top plate temperature could be as high as 550°C.

For Use With: PC-400D and PC-600D Hotplates and PC-420D and PC-620D Stirring Hotplates

  • Controls the temperature of liquids up to 200°C in vessels on top of the Pyroceram heated top
  • Increases accuracy and repeatability by controlling liquid temperature not top plate temperature
  • When in use, the controller icon on the display will be lit and the temperature control on the unit can be set from 5 to 200°C

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) PC-400D and PC-600D Hotplates and PC-420D and PC-620D Stirring Hotplates
Temperature (Metric) Ambient to 200°C
Type Temperature Controller
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.