Learn More
Delta Technologies Ltd Corning alkaline earth boro-aluminosilicate glass, 25 x 25 x 1.1 mm, Indium Tin Oxide coated one surface, Rs = 5 - 15 ohms, cut edges. Available quantities: multiples of 10, 25, 100, 250, 1,000+
SDP
Catalog No. NC1458148
Encompass
This item must be ordered in multiples of 10 per supplier requirements.ITO Coated Slides, Corning alkaline earth boro-aluminosilicate glass, 25 x 25 x 1.1 mm, Indium Tin Oxide coated one surface, resistance = 5-15 ohms/sq. Delta Technologies, Limited offers a variety of transparent, conductive indium tin oxide (ITO), flourine-doped tin oxide (FTO) coated, and uncoated products on several different substrates, custom cut to any dimension required.
Catalog No. NC1458148 Supplier Delta Technologies Ltd Supplier No. CB50IN0111
This item must be ordered in multiples of 10 per supplier requirements.ITO Coated Slides, Corning alkaline earth boro-aluminosilicate glass, 25 x 25 x 1.1 mm, Indium Tin Oxide coated one surface, resistance = 5-15 ohms/sq. Delta Technologies, Limited offers a variety of transparent, conductive indium tin oxide (ITO), flourine-doped tin oxide (FTO) coated, and uncoated products on several different substrates, custom cut to any dimension required.