Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ Graduated Disposable Polypropylene Beakers
GREENER_CHOICE

Catalog No. 0254011 Shop All Corning Products
Change view
Click to view available options
Capacity (Metric):
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
02-540-11 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-540-11 Supplier Corning™ Supplier No. 430180
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Manufactured from polypropylene for excellent chemical resistance. Corning™ Graduated Disposable Polypropylene Beakers serve as a beaker or storage container.

  • Flexible polypropylene bottom with snap-on polyethylene lid serves as a beaker or storage container.
  • Graduated in both milliliters and ounces.
  • Nonpyrogenic.
TRUSTED_SUSTAINABILITY

Specifications

Capacity (English) 8.45 oz.
Capacity (Metric) 250 mL
Material Polypropylene
Includes Beaker Only
Type Beaker
Sterility Non-sterile
Disposable Single-use
Graduated Yes
Certifications/Compliance Non-pyrogenic
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.