Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ Lambda Plus 8 and 12-Channel Pipettors
GREENER_CHOICE

Catalog No. p-4363605 Shop All Corning Products
Change view
Click to view available options
Volume (Metric):
1 to 10 μL
20 to 200 μL
5 to 50 μL
50 to 300 μL
Number of Channels:
8
12
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Number of Channels
07-764-706 1 to 10 μL 8
07-764-708 20 to 200 μL 8
07-764-707 5 to 50 μL 8
07-764-709 50 to 300 μL 8
07-764-710 1 to 10 μL 12
07-764-711 5 to 50 μL 12
07-764-713 50 to 300 μL 12
07-764-712 20 to 200 μL 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 07-764-706 Supplier Corning™ Supplier No. 4080
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Designed for universal fit with all common brands of pipet tips

Engineered to provide the highest levels of accuracy, reproducibility, and comfort

Available in single-, eight-and twelve-channel configurations, all Lambda Plus pipettors feature smooth plunger movement and extremely low pipetting forces. Volume can be set by turning either the thumb wheel or the plunger button, and a locking mechanism prevents accidental change. The Lambda Plus pipettors have been designed to maintain their calibration, even after repeated use and autoclaving. A key is included with each pipettor for easy in-house calibration.

Features

  • Factory calibrated to strict ISO 8655 standards-3-year warranty
  • Fully autoclavable and UV resistant
  • Designed for universal fit with all common brands of pipet tips
  • Adjustable tip ejector

Order Info

Accessory stand and rack available separately (Cat. Nos. 07-201-790 and -789).

Warranty and Services

3-Year

TRUSTED_SUSTAINABILITY

Specifications

Accuracy ±8 to ±2%
Certifications/Compliance CE
Autoclavable Autoclavable
For Use With Universal Pipette Tip
Number of Channels 8
Product Line Lambda Plus
Sterility Non-sterile
Type Pipette
Volume (Metric) 1 to 10 μL
UV Monitoring Yes
Warranty 3 Years
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.