Learn More
Corning™ Block
Catalog No. 07201786 Shop All Corning Products
Change view
Click to view available options
For Use With (Equipment):
LSE Digital Dry Bath Heater
Catalog No. For Use With (Equipment)
07-201-786 LSE Digital Dry Bath Heater
1 options
Catalog No. 07-201-786 Supplier Corning™ Supplier No. 480131
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart
Material: Stainless Steel; For Use With: LSE Digital Dry Bath Heater
The single and dual block configurations are used in the Corning LSE Digital Dry Bath Heaters.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) LSE Digital Dry Bath Heater
Material Stainless Steel
Type Block
For Use With (Application) 96-well plate or 4 slides