Learn More
Corning™ 384-well PCR Plate Block
Catalog No. 07201788 Shop All Corning Products
Change view
Click to view available options
For Use With (Equipment):
LSE Digital Dry Bath Heater
Catalog No. For Use With (Equipment)
07-201-788 LSE Digital Dry Bath Heater
1 options
This item is not returnable. View return policy
Catalog No. 07-201-788 Supplier Corning™ Supplier No. 480133
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy
Material: Stainless Steel; For Use With: LSE Digital Dry Bath Heater
The single and dual block configurations are used in the Corning LSE Digital Dry Bath Heaters.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) LSE Digital Dry Bath Heater
Material Stainless Steel
Type Block
For Use With (Application) 384-well PCR plate
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov