Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ Osteo Assay Surface Microplates

Catalog No. 09762102 Shop All Corning Products
Change view
Click to view available options
Volume (Metric) Working:
75 to 200 μL
Packaging:
1/Pk.; 2/Cs.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Working Packaging
09-762-102 75 to 200 μL 1/Pk.; 2/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 09-762-102 Supplier Corning™ Supplier No. 3989

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Capable of supporting the functional properties of osteogenic cells. Corning™ Osteo Assay Surface delivers lot-to-lot consistency, translating to reliable and reproducible results in bone cell assays. The surface also offers a consistent and defined alternative to preparing dentine or bone slices, reducing the variability in assay systems and resulting in more predictable assay readouts.

  • Individual alphanumerical codes for well identification, flat bottoms
  • Treated for optimal cell attachment (except where noted)
  • Corning™ CellBIND™ surface is a novel cell culture treatment that increases surface wettability for more even and consistent cell attachment.
  • Ultra-Low Attachment surface plates feature a covalently bound hydrogel layer that minimizes cell attachment, protein absorption, and cellular activation.
  • Corning Osteo Assay surface is an inorganic crystalline coating, creating a surface that mimics in vivo bone-like, for in vitro bone cell culture and assays.
  • Sterile
  • Nonpyrogenic
TRUSTED_SUSTAINABILITY

Specifications

Lid With Lid
Array 8 x 12
Packaging 1/Pk.; 2/Cs.
Color Clear
Product Type 96-well Osteo Assay Surface Multiwell Microplate
Material Polystyrene
Volume (Metric) Well 360 μL
No. of Wells 96
Volume (Metric) Working 75 to 200 μL
Sterility Sterile
Surface Treatment Osteo Assay
For Use With (Application) Osteoblast and osteoclast differentiation and functional studies
No. per Case 2/Cs.
No. per Pack 1/Pk.
Well Color Clear
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.