Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ PYREX™ Polyethylene Standard Taper Stoppers
GREENER_CHOICE

Catalog No. C762422 Shop All Corning Products
Change view
Click to view available options
Stopper Number:
9
13
16
19
22
27
38
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Stopper Number
C7624-22 22
C7624-13 13
C7624-27 27
C7624-9 9
14-629-46A 16
C7624-38 38
14-629-46B 19
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. C7624-22 Supplier Corning™ Supplier No. 762422
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Improved design fully protects neck in event of accidental tipping

These blue and white stoppers are made with closed bottoms, of linear high-density polyethylene to conform to standard taper stopper dimensions. The three unit rings on the base provide an efficient seal. The improved design fully protects the neck in the event of accidental tipping. These stoppers may be used in place of the conventional standard taper flask stoppers.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Tapered Stopper
Shape Tapered
Stopper Number 22
Material HDPE
For Use With (Application) Improved design fully protects the neck in the event of accidental tipping
Color Blue, White
Description Includes: Three rings on the plug
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.