Learn More
Description
These are various sizes and types of screw caps for use with storage bottles.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Color | Red |
| Diameter (English) Inner Neck | 1.6 in. |
| Diameter (Metric) Outer Neck | 1.77 mm |
| Diameter (Metric) Inner Neck | 41 mm |
| Material | ETFE |
| No. per Case | 50/Cs. |
| No. per Pack | 50/Pk. |
| Quantity | 50 Pk. |
| Sterility | Non-sterile |
| Thread Size (Metric) | GL 45 |
| Show More |
Frequently Asked Questions (FAQs)
Proper care includes:
• Avoiding mechanical damage (e.g., scratches or chips)
• Preventing thermal shock
• Following recommended cleaning and sterilization techniques
1. Wash bottles promptly after use, or soak them in water if immediate cleaning isn’t possible
2. Sterilize bottles contaminated with biological materials before cleaning
3. Use detergent and rinse thoroughly with tap water, followed by deionized or distilled water
4. For new bottles used in cell culture, soak in a 1% hydrochloric or nitric acid solution before washing
• Do not tighten caps immediately after autoclaving to prevent vacuum breakage, except for vented caps (e.g., 1395-45LTMC)
• Replace vented caps after 3-5 autoclave cycles or if the membrane is damaged
Do not use bottle-top filter units with reagent bottles larger than 1000 mL or laboratory bottles larger than 2000 mL.
PYREX bottles larger than 2000 mL are not recommended for vacuum applications. Additionally, 1396 and 1397 series bottles are not designed for vacuum use.
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
