Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ Primaria™ Cell Culture Multiwell™ Plates
BIOPHARMAGREENER_CHOICE

Catalog No. 087724J Shop All Corning Products
Change view
Click to view available options
Well Count:
6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Well Count
08-772-4J 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 08-772-4J Supplier Corning™ Supplier No. 353846
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Features a surface treatment that is a permanent, stable modification of the polystyrene surface incorporating a mixture of anionic and cationic functional groups. Corning™ Primaria™ Cell Culture Multiwell™ Plates feature a flat bottom with low evaporation lid.

  • Unique nitrogen-containing tissue culture surface chemistry
  • Improves attachment, spreading, and growth for many primary cells or cell lines
  • Crystal-grade polystyrene modified by proprietary vacuum-gas plasma treatment process
  • Stable, permanent surface modification
  • Optically clear
  • No special storage required
  • Samples from each lot of Corning Primaria products are analyzed by Electron Scanning for Chemical Analysis (ESCA)
  • Packaged in red color-coded, peel-open, medical-style packages
  • Sterilized by gamma irradiation
  • Nonpyrogenic tested to less than 0.1 EU/mL
TRUSTED_SUSTAINABILITY

Specifications

Array 2 x 3
Closure Color Clear
Closure Included Yes
Closure Material Polystyrene
Bottom Shape Flat
Capacity (Metric) 3 mL (Well)
Color Clear
Culture Area 9.6 cm2
Material Polystyrene
Shape Rectangular
Sterility Sterile
Surface Treatment Primaria
Type Cell Culture Plate
Volume (Metric) Working 1.5 to 2.2 mL
Well Count 6
For Use With (Application) Suitable for growing a variety of primary cells and cell lines that grow poorly on conventional, negatively charged, standard tissue culture surfaces
Includes Lid
No. per Case 50/Cs.
No. per Pack 1/Pk.
Packaging Individually Wrapped
Quantity 50/Cs.
Length (Metric) 127.63 mm
Width (Metric) 85.47 mm
Height (Metric) 21.43 mm
Diameter (Metric) 35 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.