Learn More
Description
- Unique nitrogen-containing tissue culture surface chemistry
- Improves attachment, spreading, and growth for many primary cells or cell lines
- Crystal-grade polystyrene modified by proprietary vacuum-gas plasma treatment process
- Stable, permanent surface modification
- Optically clear
- No special storage required
- Samples from each lot of Corning Primaria products are analyzed by Electron Scanning for Chemical Analysis (ESCA)
- Packaged in red color-coded, peel-open, medical-style packages
- Sterilized by gamma irradiation
- Nonpyrogenic tested to less than 0.1 EU/mL
Specifications
Specifications
| Barcode | No Barcode |
| Format | Standard |
| Lid | With Lid |
| Culture Area | 0.32 cm2 |
| Array | 8 x 12 |
| Color | Clear |
| Product Type | Microplate |
| Material | Polystyrene |
| No. of Wells | 96 |
| Volume (Metric) Working | 1.5 to 2.2 mL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
