Learn More
Corning™ Reusable Gas Venting Cap for Vertical Sidearm Flasks
Catalog No. 10270710 Shop All Corning Products
Change view
Click to view available options
No. per Case:
1/Cs.
Catalog No. No. per Case
10-270-710 1/Cs.
1 options
Catalog No. 10-270-710 Supplier Corning™ Supplier No. 4519177
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart
Used to provide gases into larger spinner flasks with vertical sidearms during cell culture
  • Used to provide gases into larger spinner flasks with vertical sidearms.
  • Fittings are comprised of a PET insert with a fluorocarbon O-Ring and a polypropylene sealing cap.
  • Gas filters are PTFE, 0.2 μm porosity.
  • The 316 stainless steel tubes are held in place by modified polyphenylene oxide thermoplastic nuts with integrated ferrules.
  • The fittings are completely autoclavable.
Spare parts for sidearm fittings, see Cat. Nos. 1395-32LTC, 1395-45LTC, 1395-LTR, and 1395LTMC
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Vertical sidearm flasks
No. per Case 1/Cs.
No. per Pack 1/Pk.
Product Type Fittings
Sterility Non-sterile
WARNING: Cancer - www.P65Warnings.ca.gov