Learn More
Corning™ PuraMatrix™ Peptide Hydrogel
Catalog No. 08774551 Shop All Corning Products
Change view
Click to view available options
Quantity:
5 mL
Catalog No. Quantity
08-774-551 5 mL
1 options
Catalog No. 08-774-551 Supplier Corning™ Supplier No. 354250
Highly defined synthetic peptide assembles into fibrous 3D structure and promotes cell attachment
Corning extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Peptide hydrogel matrix
Quantity 5 mL
WARNING: Reproductive Harm - www.P65Warnings.ca.gov