Learn More
Corning™ PYREX™ VISTA™ Class A Cylinder
SCI_ED
Catalog No. S76101F Shop All Corning Products
Change view
Click to view available options
Capacity (Metric):
10 mL
50 mL
100 mL
250 mL
500 mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
S76101F 500 mL
S76101C 50 mL
S76101A 10 mL
S76101D 100 mL
S76101E 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. S76101F Supplier Corning™ Supplier No. 70022500/EMD
This item is not returnable. View return policy
Calibrated To Contain.
  • Blue enamel graduations
  • Capacity is in accordance with ASTM™ E-1272
  • 10mL size has a funnel top for easier filling
TRUSTED_SUSTAINABILITY
Specifications
Sterility Non-sterile
Certifications/Compliance ASTM E1272
Class A
Type Graduated Cylinder
Scale Color Blue
Calibration To Contain
Material Borosilicate Glass
Capacity (Metric) 500 mL
Includes Funnel Top, Pourout
Scale Single