Learn More
Corning™ PYREX™ VISTA™ Class B Volumetric Flasks with Snap Caps
SCI_ED
Catalog No. S95185A Shop All Corning Products
Change view
Click to view available options
Capacity (Metric):
25 mL
1000 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
S95185A 25 mL
S95185F 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. S95185A Supplier Corning™ Supplier No. 7058125/EMD
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
Capacity tolerances are twice those specified for Class A volumetric ware.
  • Large, blue, block letters make inscriptions easy to read
  • Sharp, permanent graduation lines
  • Snap caps are supplied with all sizes
TRUSTED_SUSTAINABILITY
Specifications
Calibration Calibrated To Contain
Closure Color Blue
Closure Material LDPE
Capacity (Metric) 25 mL
Material Glass, Borosilicate Glass
Neck Style Long
Bottom Shape Flat