Description
- Narrow mouth
- Approximate graduations in durable blue enamel
- Extra-large marking space
Order Info
Sold in packs of 12.
Compliance
Designed to ASTM™ E-1404, Type 1.
Specifications
Specifications
| Description | Blue Enamel graduations; Top style: Beaded rim |
| Capacity (Metric) | 500 mL |
| Material | Borosilicate Glass |
| Writing Surface | Extra Large Blue Marking |
| Disposable | Reusable |
| Mouth Style | Narrow |
| Sterility | Non-sterile |