Learn More
Corning™ PYREX™ VISTA™ Flat-Bottom Boiling Flasks
SCI_ED
Catalog No. S76104B Shop All Corning Products
Change view
Click to view available options
250mL; 6/Pk.; 12/Cs.
Catalog No.
S76104B 250mL; 6/Pk.; 12/Cs.
1 options
This item is not returnable. View return policy
Catalog No. S76104B Supplier Corning™ Supplier No. 70100250/EMD
This item is not returnable. View return policy
Designed for great shock resistance.
  • Full length standard taper 24/40 outer joints
  • Short necks
  • Wall thicknesses provide balance between thermal expansion and mechanical strength
Compliance
Designed to ASTM™ E-1403.
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 250 mL
Material Borosilicate Glass
Neck Style Short
Standard Taper Joint 24/40
Disposable Reusable
Bottom Shape Flat