Learn More
Corning™ Sealing Mat for 96-well Polypropylene Plates
Catalog No. 07201112 Shop All Corning Products
Change view
Click to view available options
Material:
EVA, Polymer
Catalog No. Material
07-201-112 EVA, Polymer
1 options
Catalog No. 07-201-112 Supplier Corning™ Supplier No. 3083
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart
For use with polypropylene plates (07-200-700, -701)
  • Manufactured from EVA (ethyl vinyl acetate) and polymer is compatible with DMSO
  • DNase- and RNase-free
  • Can be applied manually or with our Storage Mat Applicator
  • Nonsterile
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance DNase and RNase-free
Material EVA, Polymer
Sterility Non-sterile
No. of Wells 96
No. per Case 50/Cs.
No. per Pack 1/Pk.
Product Type Sealing Mat
Well Shape Square
Plate Compatibility 96-well Polypropylene Plate
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov