Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ Smooth 1536-Well Multiple Well Plates
GREENER_CHOICE

Catalog No. 07200382 Shop All Corning Products
Change view
Click to view available options
Color:
Black, Clear Bottom
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
07-200-382 Black, Clear Bottom
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-200-382 Supplier Corning™ Supplier No. 3835
Only null left
Add to Cart
Add to Cart
Request a Quote

Total well volume of 10 μL for round well microplates and 12.8 μL for flat bottom microplates

Corning 1536-well microplates are our highest density microplates available for high throughput screening. The microplates conform to standard microplate footprint and dimensions. These microplates are offered in solid black and white polystyrene, with round or flat bottoms, and in black-clear bottom formats.

  • Total well volume of 10 μL for round well microplates and 12.8 μL for flat bottom microplates
  • Recommended working volume up to 8 μL
  • Round well bottom for reduced air entrapment and improved CV values and Z factor
  • Raised well bottom for higher sensitivity
  • Flood reservoir on four sides to reduce instrument contamination
  • Lids are available separately. Corning lid (Cat. No. 3098) is compatible with these microplates.
TRUSTED_SUSTAINABILITY

Specifications

Format Standard
Lid Without Lid
Array 32 x 48
Color Black, Clear Bottom
For Use With (Equipment) Automated dispensers including the PerkinElmer FlexDrop™
Material Polystyrene
No. of Wells 1536
Volume (Metric) Working 8 μL
Sterility Non-sterile
Surface Treatment Non-treated
Well Shape Flat Bottom (F-well)
For Use With (Application) For Kalypsys™ systems, optimal bottom reader sensitivity
No. per Case 100/Cs.
No. per Pack 20/Pk.
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.