Learn More
Corning™ Storage Mat III
GSA_VA
Catalog No. 07200744 Shop All Corning Products
Change view
Click to view available options
Material:
EVA, Polymer
Catalog No. Material
07-200-744 EVA, Polymer
1 options
This item is not returnable. View return policy
Catalog No. 07-200-744 Supplier Corning™ Supplier No. 3080
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Fits Corning™ 96 round well plates and blocks. Corning™ Storage Mat III can be applied manually or with a storage mat applicator.
(This is the suggested replacement for 3092 and 3094)
  • Fits Corning 96 round well plates and blocks
  • Manufactured from EVA (ethyl vinyl acetate) and polymer is compatible with DMSO
  • DNase- and RNase-free
  • Can be applied manually or with our Storage Mat Applicator
  • Nonsterile
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance DNase and RNase-free
Material EVA, Polymer
Sterility Non-sterile
No. per Case 100/Cs.
No. per Pack 25/Pk.
Product Type Storage Mat III
Well Shape Round
Plate Compatibility 96-well Plate