Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ Stripwell™ Microplates
GSA_VAGREENER_CHOICE

Catalog No. 0720097 Shop All Corning Products
Change view
Click to view available options
Surface Treatment:
Cell Culture
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Surface Treatment
07-200-97 Cell Culture
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 07-200-97 Supplier Corning™ Supplier No. 9102
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Designed and treated to provide uniform binding, high optical clarity, and low background interference. Corning™ Stripwell™ Microplates provide a superior solution for in vitro diagnostic assays.

Corning™ Stripwell™ microplates may be used in a variety of laboratory assays. The flat bottom strips are designed to easily break apart and are pre-assembled in an “egg-crate” style strip holder that allows each individual well to be positioned back into the microplate once broken.

  • Stripwell microplates have 96-well flat bottom polystyrene format.
  • Low volume and standard Stripwell microplates have well volumes of 190 μL and 360 μL, respectively.
  • 1 x 8 strips are designed to fit only one way into the strip holder, eliminating the chance of misorientation.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Format Strip Plate
Lid With Lid
Culture Area 0.32 cm2
Array 8 x 12
Binding Type TC-Treated
Color Clear
Material Polystyrene
No. of Wells 96
Volume (Metric) Working 75 to 200 μL
Sterility Sterile
Surface Treatment Cell Culture
Total Volume (Metric) Well 360 μL
Well Shape Round, Flat Bottom (F-well)
For Use With (Application) In-vitro diagnostic assays
No. per Case 50/Cs.
No. per Pack 1/Pk.
Quantity 50/Cs.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.