Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ Stripwell™ Microplates
BIOPHARMAGREENER_CHOICE

Catalog No. 0720032 Shop All Corning Products
Change view
Click to view available options
Color:
Clear
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
07-200-32 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 07-200-32 Supplier Corning™ Supplier No. 2593
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Designed and treated to provide uniform binding, high optical clarity, and low background interference. Corning™ Stripwell™ Microplates provide a superior solution for in vitro diagnostic assays.

Corning™ Stripwell™ microplates may be used in a variety of laboratory assays. The flat bottom strips are designed to easily break apart and are pre-assembled in an “egg-crate” style strip holder that allows each individual well to be positioned back into the microplate once broken.

  • Stripwell microplates have 96-well flat bottom polystyrene format.
  • Low volume and standard Stripwell microplates have well volumes of 190 μL and 360 μL, respectively.
  • 1 x 8 strips are designed to fit only one way into the strip holder, eliminating the chance of misorientation.
TRUSTED_SUSTAINABILITY

Specifications

Format Strip Plate
Lid Without Lid
Array 8 x 12
Binding Type Medium Binding, Egg crate style
Color Clear
Material Polystyrene
No. of Wells 96
Volume (Metric) Working 75 to 200 μL
Sterility Non-sterile
Surface Treatment Untreated
Total Volume (Metric) Well 360 μL
Well Shape Round
For Use With (Application) In-vitro diagnostic assays
No. per Case 100/Cs.
No. per Pack 25/Pk.
Quantity 100/Cs.
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.